Assay Design
Gene Transcripts
- Gene Symbol
- dig-1
- Entrez Gene ID
-
175951
- Gene Name
- Mesocentin
- Gene Aliases
-
CELE_K07E12.1
- Chromosome Location
-
Chr.III: 6746308 - 6794764 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001379776.1 |
NP_001366805.1 |
33 - 34 |
22006 |
114 |
View Full Product Details