Assay Design



Gene Transcripts

Gene Symbol
dig-1
Entrez Gene ID
175951
Gene Name
Mesocentin
Gene Aliases
CELE_K07E12.1
Chromosome Location
Chr.III: 6746308 - 6794764 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001379776.1 NP_001366805.1 8 - 9 1141 74

View Full Product Details