Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001027552.5 | NP_001022723.2 | 8 - 9 | 948 | 80 |
NM_001268011.3 | NP_001254940.1 | 11 - 12 | 1342 | 80 | |
NM_001268012.3 | NP_001254941.1 | 9 - 10 | 1120 | 80 | |
NM_001379777.2 | NP_001367791.1 | 8 - 9 | 954 | 80 | |
GenBank mRNA | AB206835.1 | - | 8 - 9 | 948 | 80 |
AB206836.1 | - | 3 - 4 | 454 | 80 |