Assay Design



Gene Transcripts

Gene Symbol
dpf-6
Entrez Gene ID
176132
Gene Name
protein-coding
Gene Aliases
CELE_F44B9.1
Chromosome Location
Chr.III: 8038239 - 8041407 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001382930.1 1 - 2 244 156
NM_171203.6 NP_741240.1 1 - 2 242 156

View Full Product Details