Assay Design



Gene Transcripts

Gene Symbol
elo-8
Entrez Gene ID
259739
Gene Name
protein-coding
Gene Aliases
CELE_Y47D3A.30
Chromosome Location
Chr.III: 11230377 - 11232164 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001268181.3 NP_001255110.1 3 - 4 298 74

View Full Product Details