Assay Design



Gene Transcripts

Gene Symbol
mdf-2
Entrez Gene ID
177046
Gene Name
protein-coding
Gene Aliases
CELE_Y69A2AR.30
Chromosome Location
Chr.IV: 2636227 - 2641125 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028392.7 NP_001023563.1 2 - 3 137 65
NM_001383055.1 1 - 2 83 65
GenBank mRNA AF239999.2 - 1 - 2 87 65

View Full Product Details