Assay Design



Gene Transcripts

Gene Symbol
ser-6
Entrez Gene ID
259560
Gene Name
protein-coding
Gene Aliases
CELE_Y54G2A.35
Chromosome Location
Chr.IV: 2960098 - 2964401 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001307333.2 NP_001294262.1 1 - 2 210 123
NM_171297.4 NP_741350.1 1 - 2 185 123

View Full Product Details