Assay Design



Gene Transcripts

Gene Symbol
lin-45
Entrez Gene ID
177436
Gene Name
protein-coding
Gene Aliases
CELE_Y73B6A.5
Chromosome Location
Chr.IV: 6742537 - 6748572 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306912.2 NP_001293841.1 11 - 12 2322 90
NM_001306913.3 NP_001293842.1 12 - 13 2388 90
NM_001306914.2 NP_001293843.1 11 - 12 2286 90
NM_001380288.1 NP_001368430.1 10 - 11 2087 90
NM_001383109.1 12 - 13 2345 90
NM_171367.8 NP_741430.3 11 - 12 2219 90
GenBank mRNA AY455928.1 - 11 - 12 2281 90
AY493413.1 - 11 - 12 2273 90
AY493414.1 - 10 - 11 2102 90

View Full Product Details