Assay Design



Gene Transcripts

Gene Symbol
lin-45
Entrez Gene ID
177436
Gene Name
protein-coding
Gene Aliases
CELE_Y73B6A.5
Chromosome Location
Chr.IV: 6742537 - 6748572 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306913.3 NP_001293842.1 2 - 3 286 123
NM_001383109.1 2 - 3 243 123
NM_171367.8 NP_741430.3 1 - 2 117 123
GenBank mRNA AY455928.1 - 1 - 2 179 123
AY493413.1 - 1 - 2 171 123

View Full Product Details