Assay Design
Gene Transcripts
- Gene Symbol
- col-56
- Entrez Gene ID
-
188271
- Gene Name
- COLlagen
- Gene Aliases
-
CELE_T08B2.2
- Chromosome Location
-
Chr.I: 6232777 - 6234280 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_059399.1 |
NP_491800.1 |
4 - 5 |
776 |
82 |
View Full Product Details