Assay Design
Gene Transcripts
- Gene Symbol
- srg-14
- Entrez Gene ID
-
184958
- Gene Name
- Serpentine receptor class gamma-14
- Gene Aliases
-
CELE_F26B1.6
- Chromosome Location
-
Chr.I: 6312750 - 6314624 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_059422.2 |
NP_491823.1 |
1 - 2 |
222 |
140 |
View Full Product Details