Assay Design



Gene Transcripts

Gene Symbol
mut-2
Entrez Gene ID
187005
Gene Name
protein-coding
Gene Aliases
CELE_K04F10.6
Chromosome Location
Chr.I: 6356614 - 6358411 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001372960.2 NP_001360009.1 1 - 2 196 114
NM_059433.8 NP_491834.1 1 - 2 195 114

View Full Product Details