Assay Design
Gene Transcripts
- Gene Symbol
- mei-2
- Entrez Gene ID
-
172374
- Gene Name
- Meiotic spindle formation protein 2
- Gene Aliases
-
CELE_F57B10.12
- Chromosome Location
-
Chr.I: 6564749 - 6565868 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_059493.7 |
NP_491894.1 |
1 - 2 |
132 |
75 |
GenBank mRNA |
AF248052.1 |
- |
1 - 2 |
114 |
75 |
View Full Product Details