Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001025856.4 | NP_001021027.1 | 1 - 2 | 95 | 71 |
NM_001025857.4 | NP_001021028.1 | 1 - 2 | 102 | 71 | |
GenBank mRNA | AF058331.1 | - | 1 - 2 | 115 | 71 |
AF520616.1 | - | 1 - 2 | 117 | 71 |