Assay Design



Gene Transcripts

Gene Symbol
smg-1
Entrez Gene ID
172418
Gene Name
protein-coding
Gene Aliases
CELE_C48B6.6
Chromosome Location
Chr.I: 6900943 - 6913579 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025882.4 NP_001021053.1 17 - 18 2685 81
NM_001374908.2 NP_001361976.1 17 - 17 2663 81
GenBank mRNA AF149821.1 - 17 - 18 2685 81

View Full Product Details