Assay Design



Gene Transcripts

Gene Symbol
smg-1
Entrez Gene ID
172418
Gene Name
protein-coding
Gene Aliases
CELE_C48B6.6
Chromosome Location
Chr.I: 6900943 - 6913579 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025882.4 NP_001021053.1 2 - 3 153 114
NM_001374908.2 NP_001361976.1 2 - 2 125 114
GenBank mRNA AF149821.1 - 2 - 3 153 114

View Full Product Details