Assay Design



Gene Transcripts

Gene Symbol
lin-61
Entrez Gene ID
172467
Gene Name
protein-coding
Gene Aliases
CELE_R06C7.7
Chromosome Location
Chr.I: 7248827 - 7251282 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001129029.3 NP_001122501.1 6 - 7 1759 77
NM_059649.6 NP_492050.3 5 - 6 1396 77

View Full Product Details