Assay Design
Gene Transcripts
- Gene Symbol
- tyr-3
- Entrez Gene ID
-
172472
- Gene Name
- Putative tyrosinase-like protein tyr-3
- Gene Aliases
-
CELE_F21C3.2
- Chromosome Location
-
Chr.I: 7274282 - 7278573 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_059654.5 |
NP_492055.2 |
10 - 11 |
1752 |
88 |
View Full Product Details