Assay Design
Gene Transcripts
- Gene Symbol
- ego-1
- Entrez Gene ID
-
172524
- Gene Name
- Enhancer of Glp-One (glp-1)
- Gene Aliases
-
CELE_F26A3.3
- Chromosome Location
-
Chr.I: 7650341 - 7656338 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_059731.6 |
NP_492132.1 |
3 - 4 |
717 |
108 |
GenBank mRNA |
AF159143.1 |
- |
3 - 4 |
715 |
108 |
View Full Product Details