Assay Design
Gene Transcripts
- Gene Symbol
- del-4
- Entrez Gene ID
-
189033
- Gene Name
- DEgenerin Like
- Gene Aliases
-
CELE_T28B8.5
- Chromosome Location
-
Chr.I: 8139465 - 8142537 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_059829.6 |
NP_492230.2 |
8 - 9 |
1081 |
74 |
View Full Product Details