Assay Design



Gene Transcripts

Gene Symbol
ceh-8
Entrez Gene ID
191617
Gene Name
protein-coding
Gene Aliases
CELE_ZK265.4
Chromosome Location
Chr.I: 8245505 - 8250948 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001318276.4 NP_001305205.1 4 - 4 439 75
NM_059845.4 NP_492246.2 3 - 4 463 75

View Full Product Details