Assay Design



Gene Transcripts

Gene Symbol
cutl-8
Entrez Gene ID
3565740
Gene Name
protein-coding
Gene Aliases
CELE_ZK265.8
Chromosome Location
Chr.I: 8270304 - 8275706 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001263792.3 NP_001250721.1 6 - 7 1250 109

View Full Product Details