Assay Design
Gene Transcripts
- Gene Symbol
- rsd-6
- Entrez Gene ID
-
172621
- Gene Name
- Tudor domain-containing protein
- Gene Aliases
-
CELE_F16D3.2
- Chromosome Location
-
Chr.I: 8358248 - 8363858 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_059868.9 |
NP_492269.2 |
5 - 6 |
1080 |
127 |
View Full Product Details