Assay Design
Gene Transcripts
- Gene Symbol
- daf-8
- Entrez Gene ID
-
187612
- Gene Name
- Dwarfin sma
- Gene Aliases
-
CELE_R05D11.1
- Chromosome Location
-
Chr.I: 8584081 - 8587505 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_059920.5 |
NP_492321.1 |
2 - 3 |
582 |
70 |
View Full Product Details