Assay Design
Gene Transcripts
- Gene Symbol
- sup-17
- Entrez Gene ID
-
172689
- Gene Name
- Disintegrin and metalloproteinase domain-containing protein 10 homolog
- Gene Aliases
-
CELE_DY3.7
- Chromosome Location
-
Chr.I: 8792957 - 8797298 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_059976.7 |
NP_492377.1 |
11 - 12 |
1857 |
127 |
GenBank mRNA |
AF024614.1 |
- |
11 - 12 |
1848 |
127 |
View Full Product Details