Assay Design



Gene Transcripts

Gene Symbol
ztf-26
Entrez Gene ID
172699
Gene Name
protein-coding
Gene Aliases
CELE_F55H12.6
Chromosome Location
Chr.I: 8853589 - 8861485 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026331.4 NP_001021502.1 5 - 6 682 83
NM_001026332.2 NP_001021503.1 3 - 4 620 83
NM_001373222.2 NP_001359708.1 6 - 7 882 83

View Full Product Details