Assay Design
Gene Transcripts
- Gene Symbol
- snf-2
- Entrez Gene ID
-
172701
- Gene Name
- Transporter
- Gene Aliases
-
CELE_F55H12.1
- Chromosome Location
-
Chr.I: 8871791 - 8875257 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_059995.5 |
NP_492396.2 |
1 - 2 |
494 |
78 |
View Full Product Details