Assay Design
Gene Transcripts
- Gene Symbol
- unc-29
- Entrez Gene ID
-
172703
- Gene Name
- Acetylcholine receptor subunit beta-type unc-29
- Gene Aliases
-
CELE_T08G11.5
- Chromosome Location
-
Chr.I: 8897610 - 8901435 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_059998.6 |
NP_492399.1 |
9 - 10 |
1014 |
74 |
View Full Product Details