Assay Design



Gene Transcripts

Gene Symbol
cutl-7
Entrez Gene ID
186153
Gene Name
ZP domain-containing protein
Gene Aliases
CELE_F53B6.6
Chromosome Location
Chr.I: 8954559 - 8957394 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001374976.1 NP_001361852.1 6 - 7 744 109

View Full Product Details