Assay Design
Gene Transcripts
- Gene Symbol
- clec-91
- Entrez Gene ID
-
172736
- Gene Name
- C-type lectin domain-containing protein 91
- Gene Aliases
-
CELE_ZK858.3
- Chromosome Location
-
Chr.I: 9126140 - 9127101 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_060047.5 |
NP_492448.1 |
1 - 2 |
197 |
73 |
View Full Product Details