Assay Design
Gene Transcripts
- Gene Symbol
- prom-1
- Entrez Gene ID
-
185001
- Gene Name
- protein-coding
- Gene Aliases
-
CELE_F26H9.1
- Chromosome Location
-
Chr.I: 9287443 - 9289770 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_060076.8 |
NP_492477.2 |
6 - 7 |
815 |
104 |
View Full Product Details