Assay Design
Gene Transcripts
- Gene Symbol
- hsp-70
- Entrez Gene ID
-
172757
- Gene Name
- Heat Shock Protein
- Gene Aliases
-
CELE_C12C8.1
- Chromosome Location
-
Chr.I: 9320325 - 9322628 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_060084.5 |
NP_492485.1 |
1 - 2 |
223 |
93 |
View Full Product Details