Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001381073.1 | NP_001366985.1 | 2 - 3 | 101 | 140 |
NM_001383633.1 | 2 - 3 | 150 | 140 | ||
GenBank mRNA | AF156887.1 | - | 2 - 3 | 126 | 140 |
Y17255.1 | - | - | 77 | 140 |