Assay Design
Gene Transcripts
- Gene Symbol
- klp-16
- Entrez Gene ID
-
172784
- Gene Name
- Kinesin motor domain-containing protein
- Gene Aliases
-
CELE_C41G7.2
- Chromosome Location
-
Chr.I: 9512452 - 9514755 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_060126.8 |
NP_492527.1 |
3 - 4 |
1447 |
73 |
GenBank mRNA |
AB033380.1 |
- |
3 - 4 |
1375 |
73 |
View Full Product Details