Assay Design



Gene Transcripts

Gene Symbol
rig-5
Entrez Gene ID
172791
Gene Name
protein-coding
Gene Aliases
CELE_C36F7.4
Chromosome Location
Chr.I: 9565389 - 9574377 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001128955.4 NP_001122427.1 1 - 2 236 91
NM_001264200.4 NP_001251129.1 1 - 2 236 91
NM_001264202.1 NP_001251131.1 1 - 2 236 91
NM_001381092.1 NP_001366862.1 1 - 2 46 91
NM_060134.4 NP_492535.1 1 - 2 236 91

View Full Product Details