Assay Design



Gene Transcripts

Gene Symbol
rig-5
Entrez Gene ID
172791
Gene Name
protein-coding
Gene Aliases
CELE_C36F7.4
Chromosome Location
Chr.I: 9565389 - 9574377 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001128955.4 NP_001122427.1 10 - 11 1286 63
NM_001264202.1 NP_001251131.1 11 - 12 1364 63
NM_001264203.2 NP_001251132.1 11 - 12 1211 63
NM_060134.4 NP_492535.1 9 - 10 1124 63

View Full Product Details