Assay Design



Gene Transcripts

Gene Symbol
rig-5
Entrez Gene ID
172791
Gene Name
protein-coding
Gene Aliases
CELE_C36F7.4
Chromosome Location
Chr.I: 9565389 - 9574377 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001128956.4 NP_001122428.1 4 - 5 411 142
NM_001264200.4 NP_001251129.1 9 - 10 1191 142
NM_001381092.1 NP_001366862.1 8 - 9 839 142

View Full Product Details