Assay Design



Gene Transcripts

Gene Symbol
hsr-9
Entrez Gene ID
3565010
Gene Name
Protein hsr-9
Gene Aliases
CELE_T05F1.6
Chromosome Location
Chr.I: 9626241 - 9630848 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026439.3 NP_001021610.1 8 - 9 3164 81

View Full Product Details