Assay Design
Gene Transcripts
- Gene Symbol
- zig-1
- Entrez Gene ID
-
192086
- Gene Name
- Zwei Ig domain protein zig-1
- Gene Aliases
-
CELE_K10C3.3
- Chromosome Location
-
Chr.I: 9847406 - 9850082 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_060207.6 |
NP_492608.2 |
3 - 4 |
355 |
79 |
GenBank mRNA |
AF456248.1 |
- |
3 - 4 |
354 |
79 |
View Full Product Details