Assay Design



Gene Transcripts

Gene Symbol
oct-1
Entrez Gene ID
172841
Gene Name
protein-coding
Gene Aliases
CELE_F52F12.1
Chromosome Location
Chr.I: 9888356 - 9892223 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026311.7 NP_001021482.1 7 - 8 782 125
NM_060216.6 NP_492617.4 8 - 9 845 125
GenBank mRNA AF110415.1 - 7 - 8 790 125

View Full Product Details