Assay Design



Gene Transcripts

Gene Symbol
mom-4
Entrez Gene ID
172842
Gene Name
Mitogen-activated protein kinase kinase kinase mom-4
Gene Aliases
CELE_F52F12.3
Chromosome Location
Chr.I: 9895678 - 9900817 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_060219.9 NP_492620.1 7 - 8 1359 74
GenBank mRNA AF143242.1 - 7 - 8 1350 74
AF145377.1 - 7 - 8 1141 74

View Full Product Details