Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001128976.3 | NP_001122448.1 | 2 - 3 | 220 | 89 |
NM_001128977.1 | NP_001122449.1 | 2 - 3 | 208 | 89 | |
GenBank mRNA | DQ907009.1 | - | 2 - 3 | 208 | 89 |
DQ907010.1 | - | 2 - 3 | 208 | 89 |