Assay Design



Gene Transcripts

Gene Symbol
eak-6
Entrez Gene ID
184324
Gene Name
protein-coding
Gene Aliases
CELE_F10G8.4
Chromosome Location
Chr.I: 10023798 - 10025838 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001128976.3 NP_001122448.1 4 - 5 659 156
NM_001128977.1 NP_001122449.1 4 - 5 647 156
GenBank mRNA DQ907009.1 - 4 - 5 647 156
DQ907010.1 - 4 - 5 647 156

View Full Product Details