Assay Design
Gene Transcripts
- Gene Symbol
- csnk-1
- Entrez Gene ID
-
172891
- Gene Name
- Casein kinase I gamma
- Gene Aliases
-
CELE_Y106G6E.6
- Chromosome Location
-
Chr.I: 10226986 - 10233386 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_060293.6 |
NP_492694.1 |
3 - 4 |
438 |
105 |
View Full Product Details