Assay Design
Gene Transcripts
- Gene Symbol
- wts-1
- Entrez Gene ID
-
172896
- Gene Name
- Serine/threonine-protein kinase WARTS homolog
- Gene Aliases
-
CELE_T20F10.1
- Chromosome Location
-
Chr.I: 10291870 - 10297927 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_060298.5 |
NP_492699.1 |
4 - 5 |
924 |
146 |
View Full Product Details