Assay Design
Gene Transcripts
- Gene Symbol
- tag-68
- Entrez Gene ID
-
172930
- Gene Name
- Dwarfin sma
- Gene Aliases
-
CELE_F37D6.6
- Chromosome Location
-
Chr.I: 10501585 - 10505949 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_060345.4 |
NP_492746.2 |
7 - 8 |
1129 |
110 |
View Full Product Details