Assay Design



Gene Transcripts

Gene Symbol
che-13
Entrez Gene ID
186607
Gene Name
protein-coding
Gene Aliases
CELE_F59C6.7
Chromosome Location
Chr.I: 10506998 - 10515586 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001264468.3 NP_001251397.1 5 - 6 556 77
NM_001264469.3 NP_001251398.1 2 - 3 215 77
GenBank mRNA AF520567.1 - 5 - 6 476 77

View Full Product Details