Assay Design



Gene Transcripts

Gene Symbol
xpo-2
Entrez Gene ID
171621
Gene Name
protein-coding
Gene Aliases
CELE_Y48G1A.5
Chromosome Location
Chr.I: 318150 - 323352 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306332.3 NP_001293261.1 1 - 2 287 70
NM_058315.6 NP_490716.1 4 - 5 2516 70

View Full Product Details