Assay Design



Gene Transcripts

Gene Symbol
haf-4
Entrez Gene ID
171639
Gene Name
protein-coding
Gene Aliases
CELE_W04C9.1
Chromosome Location
Chr.I: 489969 - 496158 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026495.5 NP_001021666.1 6 - 7 1122 64
NM_001306255.3 NP_001293184.1 5 - 6 842 64

View Full Product Details