Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001306244.3 | NP_001293173.1 | 2 - 3 | 240 | 84 |
NM_001306245.3 | NP_001293174.1 | 6 - 7 | 771 | 84 | |
NM_058373.6 | NP_490774.2 | 8 - 9 | 1238 | 84 | |
GenBank mRNA | AB015041.1 | - | 8 - 9 | 1095 | 84 |